Assay Details
| Gene Symbol: CRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030748.1 | NP_001025919.1 | 2 - 3 | 385 | 83 | |
| XM_015283756.2 | XP_015139242.1 | 2 - 3 | 462 | 83 | ||
| XM_025148182.1 | XP_025003950.1 | 2 - 3 | 773 | 83 | ||
| GenBank mRNA | AJ851667.1 | - | 2 - 3 | 385 | 83 | |
More Information
Set Membership: |
|

CRADD
Genomic Map