Assay Details
| Gene Symbol: ST13P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030757.1 | NP_001025928.1 | 2 - 3 | 236 | 62 | |
| XM_025148552.1 | XP_025004320.1 | 2 - 3 | 226 | 62 | ||
| GenBank mRNA | AJ719784.1 | - | 2 - 3 | 236 | 62 | |
More Information
Set Membership: |
|

ST13P5
Genomic Map