Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030768.1 | NP_001025939.1 | 4 - 5 | 904 | 104 | |
| XM_015285872.2 | XP_015141358.1 | 5 - 6 | 1907 | 104 | ||
| XM_015285888.2 | XP_015141374.1 | 5 - 6 | 1840 | 104 | ||
| XM_015285897.2 | XP_015141383.1 | 5 - 6 | 1052 | 104 | ||
| XM_015285901.2 | XP_015141387.1 | 3 - 4 | 1597 | 104 | ||
| XM_025148873.1 | XP_025004641.1 | 3 - 4 | 911 | 104 | ||
| GenBank mRNA | AJ721030.1 | - | 4 - 5 | 904 | 104 | |
More Information
Set Membership: |
|

BPGM
Genomic Map