Assay Details
| Gene Symbol: NAA50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030778.1 | NP_001025949.1 | 2 - 3 | 382 | 64 | |
| XM_015294532.2 | XP_015150018.1 | 2 - 3 | 392 | 64 | ||
| XM_015294536.2 | XP_015150022.1 | 2 - 3 | 342 | 64 | ||
| GenBank mRNA | AJ719306.1 | - | 2 - 3 | 382 | 64 | |
More Information
Set Membership: |
|

NAA50
Genomic Map