Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030791.1 | NP_001025962.1 | 1 - 2 | 82 | 67 | |
| XM_025149429.1 | XP_025005197.1 | 1 - 2 | 285 | 67 | ||
| XM_025149433.1 | XP_025005201.1 | 1 - 2 | 283 | 67 | ||
| GenBank mRNA | AJ851452.1 | - | 1 - 2 | 82 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025149433.1 transcripts.
Set Membership: |
|

GPR89A
Genomic Map