Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030791.1 | NP_001025962.1 | 3 - 4 | 247 | 107 | |
| XM_025149429.1 | XP_025005197.1 | 3 - 4 | 450 | 107 | ||
| XM_025149433.1 | XP_025005201.1 | 3 - 4 | 531 | 107 | ||
| GenBank mRNA | AJ851452.1 | - | 3 - 4 | 247 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025149433.1 transcripts.
Set Membership: |
|

GPR89A
Genomic Map