Assay Details
| Gene Symbol: TMEM57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030895.2 | NP_001026066.2 | 8 - 9 | 1715 | 76 | |
| XM_025142989.1 | XP_024998757.1 | 6 - 7 | 1261 | 76 | ||
| GenBank mRNA | AJ719738.1 | - | 8 - 9 | 1745 | 76 | |
| AY845022.1 | - | 8 - 9 | 1715 | 76 | ||
More Information
Set Membership: |
|

TMEM57
Genomic Map