Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030946.1 | NP_001026117.1 | 2 - 3 | 331 | 72 | |
| XM_015282921.2 | XP_015138407.1 | 2 - 3 | 419 | 72 | ||
| XM_015282923.2 | XP_015138409.1 | 2 - 3 | 768 | 72 | ||
| XM_015282925.2 | XP_015138411.1 | 2 - 3 | 423 | 72 | ||
| GenBank mRNA | AJ851440.1 | - | 2 - 3 | 331 | 72 | |
More Information
Set Membership: |
|

TP53INP1
Genomic Map