Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030946.1 | NP_001026117.1 | 3 - 4 | 692 | 73 | |
| XM_015282921.2 | XP_015138407.1 | 3 - 4 | 780 | 73 | ||
| XM_015282923.2 | XP_015138409.1 | 3 - 4 | 1129 | 73 | ||
| XM_015282924.2 | XP_015138410.1 | 3 - 4 | 687 | 73 | ||
| GenBank mRNA | AJ851440.1 | - | 3 - 4 | 692 | 73 | |
More Information
Set Membership: |
|

TP53INP1
Genomic Map