Assay Details
| Gene Symbol: SQLE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194927.1 | NP_001181856.1 | 10 - 11 | 2036 | 67 | |
| GenBank mRNA | AJ720030.1 | - | 10 - 11 | 2036 | 67 | |
More Information
Set Membership: |
|

SQLE
Genomic Map