Assay Details
| Gene Symbol: ANKMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030979.1 | NP_001026150.1 | 1 - 2 | 149 | 58 | |
| GenBank mRNA | AJ719452.1 | - | 1 - 2 | 149 | 58 | |
More Information
Set Membership: |
|

ANKMY2
Genomic Map