Assay Details
| Gene Symbol: NUP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030984.1 | NP_001026155.1 | 2 - 3 | 421 | 71 | |
| XM_015281442.2 | XP_015136928.2 | 2 - 3 | 482 | 71 | ||
| GenBank mRNA | AJ720962.1 | - | 2 - 3 | 421 | 71 | |
More Information
Set Membership: |
|

NUP42
Genomic Map