Assay Details
| Gene Symbol: KDSR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031002.1 | NP_001026173.1 | 6 - 7 | 768 | 77 | |
| XM_015282176.2 | XP_015137662.1 | 6 - 7 | 77 | |||
| XM_015282177.2 | XP_015137663.1 | 6 - 7 | 77 | |||
| GenBank mRNA | AJ719938.1 | - | 6 - 7 | 768 | 77 | |
More Information
Set Membership: |
|

KDSR
Genomic Map