Assay Details
| Gene Symbol: TNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031166.2 | NP_001026337.2 | 3 - 4 | 897 | 96 | |
| XM_015285792.2 | XP_015141278.1 | 3 - 4 | 3472 | 96 | ||
| GenBank mRNA | AJ851742.1 | - | 3 - 4 | 903 | 96 | |
More Information
Set Membership: |
|

TNIP2
Genomic Map