Assay Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031180.1 | NP_001026351.1 | 9 - 10 | 1482 | 72 | |
| XM_015286385.1 | XP_015141871.1 | 9 - 10 | 1283 | 72 | ||
| GenBank mRNA | AJ851388.1 | - | 9 - 10 | 1482 | 72 | |
More Information
Set Membership: |
|

SWAP70
Genomic Map