Assay Details
| Gene Symbol: BTF3L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031285.1 | NP_001026456.1 | 2 - 3 | 171 | 65 | |
| GenBank mRNA | AJ720471.1 | - | 2 - 3 | 171 | 65 | |
More Information
Set Membership: |
|

BTF3L4
Genomic Map