Assay Details
| Gene Symbol: SERBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031293.1 | NP_001026464.1 | 8 - 9 | 1320 | 78 | |
| XM_015291044.2 | XP_015146530.1 | 9 - 10 | 1407 | 78 | ||
| XM_015291045.2 | XP_015146531.1 | 9 - 10 | 1383 | 78 | ||
| XM_015291046.2 | XP_015146532.1 | 8 - 9 | 1358 | 78 | ||
| GenBank mRNA | AJ851638.1 | - | 8 - 9 | 1320 | 78 | |
More Information
Set Membership: |
|

SERBP1
Genomic Map