Assay Details
| Gene Symbol: KLHL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031304.2 | NP_001026475.1 | 3 - 4 | 3539 | 69 | |
| GenBank mRNA | AJ851506.1 | - | 3 - 4 | 3540 | 69 | |
More Information
Set Membership: |
|

KLHL24
Genomic Map