Assay Details
| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031316.1 | NP_001026487.1 | 7 - 8 | 743 | 57 | |
| XM_015291407.2 | XP_015146893.1 | 7 - 8 | 779 | 57 | ||
| GenBank mRNA | AJ720614.1 | - | 7 - 8 | 743 | 57 | |
More Information
Set Membership: |
|

FYTTD1
Genomic Map