Assay Details
| Gene Symbol: ORMDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031325.1 | NP_001026496.1 | 2 - 3 | 240 | 60 | |
| XM_025145284.1 | XP_025001052.1 | 2 - 3 | 215 | 60 | ||
| GenBank mRNA | AJ720694.1 | - | 2 - 3 | 240 | 60 | |
More Information
Set Membership: |
|

ORMDL2
Genomic Map