Assay Details
| Gene Symbol: CLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031326.1 | NP_001026497.1 | 2 - 3 | 324 | 93 | |
| XM_015298611.2 | XP_015154097.1 | 2 - 3 | 300 | 93 | ||
| XM_025143396.1 | XP_024999164.1 | 2 - 3 | 300 | 93 | ||
| XM_025143397.1 | XP_024999165.1 | 2 - 3 | 318 | 93 | ||
| XM_025143398.1 | XP_024999166.1 | 2 - 3 | 300 | 93 | ||
| GenBank mRNA | AJ720552.1 | - | 2 - 3 | 324 | 93 | |
More Information
Set Membership: |
|

CLK2
Genomic Map