Assay Details
| Gene Symbol: C10orf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031344.1 | NP_001026515.1 | 1 - 2 | 1351 | 59 | |
| XM_015288692.2 | XP_015144178.1 | 1 - 2 | 1372 | 59 | ||
| GenBank mRNA | AJ720673.1 | - | 1 - 2 | 1351 | 59 | |
More Information
Set Membership: |
|

C10orf2
Genomic Map