Assay Details
| Gene Symbol: RBM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031347.1 | NP_001026518.1 | 3 - 4 | 272 | 101 | |
| XM_015299933.2 | XP_015155419.1 | 3 - 4 | 272 | 101 | ||
| XM_015299934.2 | XP_015155420.1 | 3 - 4 | 2111 | 101 | ||
| XM_015299935.2 | XP_015155421.1 | 3 - 4 | 301 | 101 | ||
| XM_015299936.2 | XP_015155422.1 | 3 - 4 | 305 | 101 | ||
| XM_015299937.2 | XP_015155423.1 | 3 - 4 | 272 | 101 | ||
| XM_015299938.2 | XP_015155424.1 | 3 - 4 | 274 | 101 | ||
| GenBank mRNA | AJ719636.1 | - | 3 - 4 | 272 | 101 | |
| DQ126684.1 | - | 2 - 3 | 211 | 101 | ||
| KC905084.1 | - | 2 - 3 | 211 | 101 | ||
More Information
Set Membership: |
|

RBM3
Genomic Map