Assay Details
| Gene Symbol: VPS33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031348.1 | NP_001026519.1 | 4 - 5 | 339 | 65 | |
| XM_015292119.2 | XP_015147605.1 | 4 - 5 | 325 | 65 | ||
| GenBank mRNA | AJ719863.1 | - | 4 - 5 | 339 | 65 | |
| CR385810.1 | - | 4 - 5 | 310 | 65 | ||
More Information
Set Membership: |
|

VPS33B
Genomic Map