Assay Details
| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031361.1 | NP_001026532.1 | 9 - 10 | 1284 | 55 | |
| XM_015299512.2 | XP_015154998.1 | 7 - 8 | 2591 | 55 | ||
| XM_015299513.2 | XP_015154999.1 | 9 - 10 | 3824 | 55 | ||
| XM_015299514.2 | XP_015155000.1 | 8 - 9 | 2477 | 55 | ||
| XM_015299515.2 | XP_015155001.1 | 8 - 9 | 1433 | 55 | ||
| XM_025143982.1 | XP_024999750.1 | 8 - 9 | 4221 | 55 | ||
| XM_025143983.1 | XP_024999751.1 | 8 - 9 | 4224 | 55 | ||
| XM_025143984.1 | XP_024999752.1 | 9 - 10 | 3311 | 55 | ||
| XM_025143985.1 | XP_024999753.1 | 10 - 11 | 3713 | 55 | ||
| GenBank mRNA | AJ721093.1 | - | 9 - 10 | 1284 | 55 | |
More Information
Set Membership: |
|

GSDMA
Genomic Map