Assay Details
| Gene Symbol: SRSF7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031368.1 | NP_001026539.1 | 6 - 7 | 688 | 88 | |
| GenBank mRNA | AJ719404.1 | - | 6 - 7 | 688 | 88 | |
| CR385735.1 | - | 3 - 4 | 564 | 88 | ||
More Information
Set Membership: |
|

SRSF7L
Genomic Map