Assay Details
| Gene Symbol: SRSF7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031368.1 | NP_001026539.1 | 7 - 8 | 728 | 81 | |
| XM_015283521.2 | XP_015139007.1 | 8 - 9 | 773 | 81 | ||
| GenBank mRNA | AJ719404.1 | - | 7 - 8 | 728 | 81 | |
| CR385735.1 | - | 4 - 5 | 604 | 81 | ||
More Information
Set Membership: |
|

SRSF7L
Genomic Map