Assay Details
| Gene Symbol: SUV39H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031370.2 | NP_001026541.1 | 5 - 6 | 1136 | 75 | |
| GenBank mRNA | AJ851535.1 | - | 5 - 6 | 1137 | 75 | |
More Information
Set Membership: |
|

SUV39H2
Genomic Map