Assay Details
| Gene Symbol: H2AFZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031374.1 | NP_001026545.1 | 3 - 4 | 350 | 71 | |
| GenBank mRNA | AJ719448.1 | - | 3 - 4 | 350 | 71 | |
More Information
Set Membership: |
|

H2AFZ
Genomic Map