Assay Details
| Gene Symbol: PGM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031383.1 | NP_001026554.1 | 1 - 2 | 74 | 130 | |
| XM_025149745.1 | XP_025005513.1 | 1 - 2 | 335 | 130 | ||
| GenBank mRNA | AJ721042.1 | - | 1 - 2 | 74 | 130 | |
More Information
Set Membership: |
|

PGM2
Genomic Map