Assay Details
| Gene Symbol: CIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031386.1 | NP_001026557.1 | 8 - 9 | 635 | 62 | |
| GenBank mRNA | AJ720981.1 | - | 8 - 9 | 635 | 62 | |
More Information
Set Membership: |
|

CIR1
Genomic Map