Assay Details
| Gene Symbol: RFXANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031389.1 | NP_001026560.1 | 3 - 4 | 422 | 80 | |
| XM_015300077.2 | XP_015155563.2 | 3 - 4 | 451 | 80 | ||
| GenBank mRNA | AJ720002.1 | - | 3 - 4 | 422 | 80 | |
| CR406288.1 | - | 1 - 2 | 36 | 80 | ||
More Information
Set Membership: |
|

RFXANK
Genomic Map