Assay Details
| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031393.1 | NP_001026564.1 | 2 - 3 | 282 | 64 | |
| GenBank mRNA | AJ721023.1 | - | 2 - 3 | 282 | 64 | |
| CR352610.1 | - | 1 - 2 | 136 | 64 | ||
More Information
Set Membership: |
|

KNSTRN
Genomic Map