Assay Details
| Gene Symbol: ALG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031394.1 | NP_001026565.1 | 1 - 2 | 197 | 117 | |
| GenBank mRNA | AJ720171.1 | - | 1 - 2 | 197 | 117 | |
| CR352406.1 | - | 1 - 2 | 128 | 117 | ||
More Information
Set Membership: |
|

ALG10
Genomic Map