Assay Details
| Gene Symbol: DYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031398.1 | NP_001026569.1 | 8 - 9 | 935 | 63 | |
| XM_015277288.2 | XP_015132774.1 | 8 - 9 | 955 | 63 | ||
| XM_015277289.2 | XP_015132775.1 | 9 - 10 | 1084 | 63 | ||
| GenBank mRNA | AJ719621.1 | - | 8 - 9 | 935 | 63 | |
More Information
Set Membership: |
|

DYM
Genomic Map