Assay Details
| Gene Symbol: EFHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031773.1 | NP_001026943.1 | 3 - 4 | 676 | 87 | |
| XM_025153331.1 | XP_025009099.1 | 3 - 4 | 384 | 87 | ||
| GenBank mRNA | AY971655.1 | - | 3 - 4 | 676 | 87 | |
| BX933977.2 | - | 3 - 4 | 579 | 87 | ||
| BX934902.1 | - | 2 - 3 | 279 | 87 | ||
More Information
Set Membership: |
|

EFHD1
Genomic Map