Assay Details
| Gene Symbol: CNIH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033006.1 | NP_001028178.1 | 1 - 2 | 106 | 63 | |
| GenBank mRNA | AB232678.1 | - | 1 - 2 | 106 | 63 | |
| BX929843.1 | - | 1 - 2 | 108 | 63 | ||
| BX931023.1 | - | 1 - 2 | 157 | 63 | ||
| BX933003.1 | - | 1 - 2 | 87 | 63 | ||
| BX936003.1 | - | 1 - 2 | 92 | 63 | ||
More Information
Set Membership: |
|

CNIH1
Genomic Map