Assay Details
| Gene Symbol: TNFRSF11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033641.1 | NP_001028813.1 | 2 - 3 | 410 | 67 | |
| XM_015283019.2 | XP_015138505.1 | 2 - 3 | 634 | 67 | ||
| GenBank mRNA | CR352482.1 | - | 2 - 3 | 478 | 67 | |
| DQ098013.1 | - | 2 - 3 | 410 | 67 | ||
More Information
Set Membership: |
|

TNFRSF11B
Genomic Map