Assay Details
| Gene Symbol: C4BPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033643.1 | NP_001028815.1 | 500 | 104 | ||
| GenBank mRNA | AB109024.1 | - | 500 | 104 | ||
| BX934798.2 | - | 206 | 104 | |||
More Information
Set Membership: |
|

C4BPG
Genomic Map