Assay Details
| Gene Symbol: C4BPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033643.1 | NP_001028815.1 | 1155 | 92 | ||
| GenBank mRNA | AB109024.1 | - | 1155 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4BPG| Gene Symbol: C4BPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033643.1 | NP_001028815.1 | 1155 | 92 | ||
| GenBank mRNA | AB109024.1 | - | 1155 | 92 | ||