Assay Details
| Gene Symbol: CETP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034814.2 | NP_001029986.2 | 10 - 11 | 1029 | 72 | |
| XM_025154152.1 | XP_025009920.1 | 17 - 18 | 5313 | 72 | ||
| GenBank mRNA | AB205527.1 | - | 10 - 11 | 1055 | 72 | |
More Information
Set Membership: |
|

CETP
Genomic Map