Assay Details
| Gene Symbol: PROCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034820.1 | NP_001029992.1 | 3 - 4 | 579 | 125 | |
| GenBank mRNA | AB204800.1 | - | 3 - 4 | 579 | 125 | |
More Information
Set Membership: |
|

PROCR
Genomic Map