Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034827.2 | NP_001029999.1 | 7 - 8 | 585 | 71 | |
| XM_015273811.2 | XP_015129297.2 | 9 - 10 | 749 | 71 | ||
| XM_015273812.2 | XP_015129298.2 | 8 - 9 | 790 | 71 | ||
| GenBank mRNA | DQ133597.1 | - | 8 - 9 | 691 | 71 | |
| DQ133598.1 | - | 7 - 8 | 587 | 71 | ||
More Information
Set Membership: |
|

PRKAG1
Genomic Map