Assay Details
| Gene Symbol: DNASE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037838.1 | NP_001032927.1 | 3 - 4 | 394 | 67 | |
| XM_015290817.2 | XP_015146303.1 | 4 - 5 | 533 | 67 | ||
| GenBank mRNA | CR386219.1 | - | 3 - 4 | 394 | 67 | |
| DQ272298.1 | - | 3 - 4 | 394 | 67 | ||
More Information
Set Membership: |
|

DNASE2B
Genomic Map