Assay Details
| Gene Symbol: CD86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037839.1 | NP_001032928.1 | 4 - 5 | 736 | 93 | |
| XM_015293509.2 | XP_015148995.1 | 3 - 4 | 1774 | 93 | ||
| GenBank mRNA | AM050135.1 | - | 4 - 5 | 736 | 93 | |
| EF554724.1 | - | 3 - 4 | 707 | 93 | ||
More Information
Set Membership: |
|

CD86
Genomic Map