Assay Details
| Gene Symbol: CNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038652.1 | NP_001033741.1 | 1 | 1447 | 74 | |
| XM_015284635.2 | XP_015140121.1 | 2 | 1586 | 74 | ||
| XM_015284636.2 | XP_015140122.1 | 2 | 1626 | 74 | ||
| XM_015284638.2 | XP_015140124.1 | 3 | 2935 | 74 | ||
More Information
Set Membership: |
|

CNR1
Genomic Map