Assay Details
| Gene Symbol: SFTPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039166.1 | NP_001034255.1 | 1 - 2 | 173 | 108 | |
| XM_025151510.1 | XP_025007278.1 | 2 - 3 | 254 | 108 | ||
| GenBank mRNA | CR391234.1 | - | 1 - 2 | 172 | 108 | |
| DQ129667.1 | - | 1 - 2 | 173 | 108 | ||
More Information
Set Membership: |
|

SFTPA1
Genomic Map