Assay Details
| Gene Symbol: CDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039258.2 | NP_001034347.2 | 2 - 3 | 284 | 130 | |
| XM_015292581.2 | XP_015148067.1 | 2 - 3 | 295 | 130 | ||
| GenBank mRNA | AJ446009.1 | - | 2 - 3 | 285 | 130 | |
| M16260.1 | - | 2 - 3 | 234 | 130 | ||
More Information
Set Membership: |
|

CDH1
Genomic Map