Assay Details
| Gene Symbol: SRPRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039277.1 | NP_001034366.1 | 13 - 14 | 1848 | 67 | |
| GenBank mRNA | AJ851760.1 | - | 13 - 14 | 1848 | 67 | |
More Information
Set Membership: |
|

SRPRA
Genomic Map