Assay Details
| Gene Symbol: ACP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039291.1 | NP_001034380.1 | 5 - 6 | 422 | 92 | |
| XM_015284838.2 | XP_015140324.1 | 5 - 6 | 438 | 92 | ||
| GenBank mRNA | AJ720119.1 | - | 5 - 6 | 422 | 92 | |
More Information
Set Membership: |
|

ACP1
Genomic Map